2-Chlor-3-cyanpyrazin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-chloropyrazine-2-carbonitrile
OR3400-5G
APOSOR3400-5GEA
21.9
EUR
APOSOR3400-5G
2-Chlor-3-cyanpyrazin 98%
2-Chloro-3-cyanopyrazine
Formel:
C₅H₂ClN₃ Molecular Weight: 139,5400 g/mol Schmelzpunkt: 38…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00219653 CAS-Nummer: 55557-52-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...