(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-3,3'-Bis(3,5-dimethylphenyl)-[1,1'-binaphthalene]-2,2'-diol

OR340001-250MG
APOSOR340001-250MGEA 172 EUR
APOSOR340001-250MG
(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol
(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol
Formel: C₃₆H₃₀O₂
Molecular Weight: 494,622 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11617327
CAS-Nummer: 435327-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR