(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binaphthalene]-2,2'-diol

OR340006-250MG
APOSOR340006-250MGEA 346 EUR
APOSOR340006-250MG
(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol
(R)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol
Formel: C₅₀H₅₈O₂
Molecular Weight: 690,994 g/mol
Schmelzpunkt: 250…262 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD11045378
CAS-Nummer: 247123-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR