(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol

Lieferant: Apollo Scientific

OR340012-250MG
APOSOR340012-250MGEA 101 EUR
APOSOR340012-250MG
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
Formel: C₃₂H₂₀N₂O₆
Lagertemperatur: Gefrierschrank
MDL: MFCD28139323
CAS-Nummer: 791616-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR