(S)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol

Lieferant: Apollo Scientific
Achtung

OR340013-250MG
APOSOR340013-250MGEA 69.6 EUR
APOSOR340013-250MG
(S)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
(S)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
Formel: C₃₂H₂₀N₂O₆
Molecular Weight: 528,511 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28139323
CAS-Nummer: 945852-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR