(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Lieferant: Apollo Scientific
OR340015-250MG
APOSOR340015-250MGEA
200
EUR
APOSOR340015-250MG
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Formel:
C₄₈H₃₀O₂ Schmelzpunkt: 239…242 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09027270 CAS-Nummer: 361342-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...