(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific

Synonyme: (11bR)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

OR340018-250MG
APOSOR340018-250MGEA 166 EUR
APOSOR340018-250MG
(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel: C₃₆H₂₉O₄P
Molecular Weight: 556,587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27952571
CAS-Nummer: 861909-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR