(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Lieferant: Apollo Scientific
Synonyme:
(11bS)-2,6-Bis(3,5-dimethylphenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
OR340019-250MG
APOSOR340019-250MGEA
151
EUR
APOSOR340019-250MG
(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(S)-3,3'-Bis[3,5-dimethylphenyl]-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel:
C₃₆H₂₉O₄P Molecular Weight: 556,587 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27952571 CAS-Nummer: 1170736-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...