(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-3,3'-Bis(2,4,6-triisopropylphenyl)-1,1'-bi-2-naphthol cyclic monophosphate, (R)-TRIP, (11bR)-4-Hydroxy-2,6-bis[2,4,6-tris(1-methylethyl)phenyl]dinaphtho[2,1-d:1',2'-f]-1,3,2-dioxaphosphepin 4-oxide monophosphate, (11bR)-4-Hydroxy-2,6-bis(2,4,6-triisopropylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

OR340024-250MG
APOSOR340024-250MGEA 97.3 EUR
APOSOR340024-250MG
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis(2,4,6-trisopropylphenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel: C₅₀H₅₇O₄P
Molecular Weight: 752,959 g/mol
Schmelzpunkt: 210…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10567009
CAS-Nummer: 791616-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR