(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific
Achtung

Synonyme: (11bS)-4-Hydroxy-2,6-bis(4-nitrophenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

OR340031-250MG
APOSOR340031-250MGEA 177 EUR
APOSOR340031-250MG
(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel: C₃₂H₁₉N₂O₈P
Molecular Weight: 590.476 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08274951
CAS-Nummer: 878111-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR