(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Lieferant: Apollo Scientific

Synonyme: (11bR)-2,6-Di(anthracen-9-yl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

OR340032-250MG
APOSOR340032-250MGEA 172 EUR
APOSOR340032-250MG
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Formel: C₄₈H₂₉O₄P
Molecular Weight: 700.715 g/mol
Schmelzpunkt: 260…274 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD11045406
CAS-Nummer: 361342-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR