N'-Hydroxy-2-pyrazinecarboximidamide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR3401-250MG
APOSOR3401-250MGEA
21.9
EUR
APOSOR3401-250MG
N'-Hydroxy-2-pyrazinecarboximidamide 97%
N'-Hydroxy-2-pyrazinecarboximidamide
Formel:
C₅H₆N₄O Schmelzpunkt: 180…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258397 CAS-Nummer: 51285-05-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...