1,4-Benzodioxan-2-carbonsäure 98%

Lieferant: Apollo Scientific

OR3409-100G
APOSOR3409-100GEA 89.9 EUR
APOSOR3409-100G
1,4-Benzodioxan-2-carbonsäure 98%
1,4-Benzodioxan-2-carbonsäure
Formel: C₉H₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00084888
CAS-Nummer: 3663-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR