2,3-Dihydrobenzo[b]furan-5-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydrobenzo[b]furan-5-carboxylicacid

OR3414-1G
APOSOR3414-1GEA 31.9 EUR
APOSOR3414-1G
2,3-Dihydrobenzo[b]furan-5-carbonsäure 98%
2,3-Dihydrobenzo[b]furan-5-carbonsäure
Formel: C₉H₈O₃
Molecular Weight: 164,1580 g/mol
Schmelzpunkt: 194…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085012
CAS-Nummer: 76429-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR