6-Ethoxy-2-naphthalinboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Ethoxynaphthalen-2-yl)boronic acid
OR3425-1G
APOSOR3425-1GEA
21.9
EUR
APOSOR3425-1G
6-Ethoxy-2-naphthalinboronsäure 98%
6-Ethoxy-2-naphthaleneboronic acid
Formel:
C₁₂H₁₃BO₃ Molecular Weight: 216,04 g/mol Schmelzpunkt: 252…255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427009 CAS-Nummer: 352525-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...