2-(Pyrrolidin-1-ylsulphonyl)benzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

OR3438-1G
APOSOR3438-1GEA 0 EUR
APOSOR3438-1G
2-(Pyrrolidin-1-ylsulphonyl)benzeneboronic acid 97%
2-(Pyrrolidin-1-ylsulphonyl)benzeneboronic acid
Formel: C₁₀H₁₄BNO₄S
Schmelzpunkt: 124…126 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689515
CAS-Nummer: 913835-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR