4-(1H-Pyrrol-1-yl)benzoic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR3439-1G
APOSOR3439-1GEA 143 EUR
APOSOR3439-1G
4-(1H-Pyrrol-1-yl)benzoic acid 95+%
4-(1H-Pyrrol-1-yl)benzoic acid
Formel: C₁₁H₉NO₂
Schmelzpunkt: 286…289 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082569
CAS-Nummer: 22106-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR