4-Chlor-2-formylphenylboronsäure 98%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-formylbenzeneboronic acid
OR3443-1G
APOSOR3443-1GEA
26.1
EUR
APOSOR3443-1G
4-Chlor-2-formylphenylboronsäure 98%
4-Chlor-2-formylphenylboronsäure
Formel:
C₇H₆BClO₃ Molecular Weight: 184,39 g/mol |
MDL:
MFCD08689492 CAS-Nummer: 913835-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...