3-Formyl-2-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3445-1G
APOSOR3445-1GEA
34.8
EUR
APOSOR3445-1G
3-Formyl-2-methoxyphenylboronsäure 98%
3-Formyl-2-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179,9700 g/mol Schmelzpunkt: 122…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689486 CAS-Nummer: 480424-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...