4-[(Benzylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Benzyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Benzylureido)benzeneboronic acid, pinacol ester
OR3448-1G
APOSOR3448-1GEA
102
EUR
APOSOR3448-1G
4-[(Benzylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
4-[(Benzylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Formel:
C₂₀H₂₅BN₂O₃ Molecular Weight: 352,235 g/mol Schmelzpunkt: 204…207 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689513 CAS-Nummer: 874290-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...