2-Methoxy-4-nitrobenzonitril 99%

Lieferant: Apollo Scientific
Achtung

OR345004-1G OR345004-5G
APOSOR345004-1GEA 50.9 EUR
APOSOR345004-1G APOSOR345004-5G
2-Methoxy-4-nitrobenzonitril 99%
2-Methoxy-4-nitrobenzonitril
Formel: C₈H₆N₂O₃
Molecular Weight: 178.15 g/mol
Schmelzpunkt: 177…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093781
CAS-Nummer: 101084-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR