4-Cyano-3-iodopyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR345006-1G
OR345006-250MG
APOSOR345006-1GEA
319
EUR
APOSOR345006-1G
APOSOR345006-250MG
4-Cyano-3-iodopyridine 98%
4-Cyano-3-iodopyridine
Formel:
C₆H₃IN₂ Schmelzpunkt: 119…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08729285 CAS-Nummer: 10386-28-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...