3,4-Dibrompyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR345020-100G
OR345020-25G
OR345020-5G
APOSOR345020-100GEA
282
EUR
APOSOR345020-100G
APOSOR345020-25G
APOSOR345020-5G
3,4-Dibrompyridin
3,4-Dibromopyridine
Formel:
C₅H₃Br₂N Schmelzpunkt: 71…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234016 CAS-Nummer: 13534-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...