3-Brom-4-hydroxybenzoesäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR345023-5G
APOSOR345023-5GEA
21.9
EUR
APOSOR345023-5G
3-Brom-4-hydroxybenzoesäure
3-Bromo-4-hydroxybenzoic acid
Formel:
C₇H₅BrO₃ Molecular Weight: 217,02 g/mol Schmelzpunkt: 155…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017547 CAS-Nummer: 14348-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...