4-Methyl-N3-[4-(3-pyridinyl)-2-pyrimidinyl]-1,3-benzoldiamin 98%

Lieferant: Apollo Scientific

Synonyme: 4-(3-pyridyl)-2-pyrimidineamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine, N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(5-Amino-2-methylanilino)-4-(3-pyridyl)pyrimidine

OR345028-100G
APOSOR345028-100GEA 88.6 EUR
APOSOR345028-100G
4-Methyl-N3-[4-(3-pyridinyl)-2-pyrimidinyl]-1,3-benzoldiamin 98%
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine
Formel: C₁₆H₁₅N₅
Molecular Weight: 277,3200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09028125
CAS-Nummer: 152460-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR