2-Amino-6-methylpyridin-3-ol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-hydroxy-6-methylpyridine
OR345045-1G
APOSOR345045-1GEA
155
EUR
APOSOR345045-1G
2-Amino-6-methylpyridin-3-ol 97%
2-Amino-6-methylpyridin-3-ol
Formel:
C₆H₈N₂O Molecular Weight: 124,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839245 CAS-Nummer: 20348-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...