4,5-Dimethoxy-2-methylbenzoic acid 97%

Lieferant: Apollo Scientific

OR345046-1G
APOSOR345046-1GEA 163 EUR
APOSOR345046-1G
4,5-Dimethoxy-2-methylbenzoic acid 97%
4,5-Dimethoxy-2-methylbenzoic acid
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00029889
CAS-Nummer: 20736-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR