4-(2-Chloroethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR345048-25G OR345048-5G
APOSOR345048-25GEA 0 EUR
APOSOR345048-25G APOSOR345048-5G
4-(2-Chloroethyl)benzoic acid
4-(2-Chloroethyl)benzoic acid
Formel: C₉H₉ClO₂
Schmelzpunkt: 187…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013995
CAS-Nummer: 20849-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR