2-Chlor-4-methoxybenzoesäure
Lieferant: Apollo Scientific
Synonyme:
2-Chlor-p-anissäure
OR345051-25G
APOSOR345051-25GEA
115
EUR
APOSOR345051-25G
2-Chlor-4-methoxybenzoesäure
2-Chloro-4-methoxybenzoic acid
Formel:
C₈H₇ClO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085943 CAS-Nummer: 21971-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...