(±)-4-Brom-α-methylbenzylamin 98%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine

OR345056-25G
APOSOR345056-25GEA 69.6 EUR
APOSOR345056-25G
(±)-4-Brom-α-methylbenzylamin 98%
(±)-4-Brom-α-methylbenzylamin
Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025548
CAS-Nummer: 24358-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR