4-[(4-Chloro-2-pyrimidinyl)amino]benzonitrile 97%
Lieferant: Apollo Scientific
Synonyme:
4-((4-Chloropyrimidin-2-yl)amino)benzonitrile
OR345057-1G
APOSOR345057-1GEA
81.2
EUR
APOSOR345057-1G
4-[(4-Chloro-2-pyrimidinyl)amino]benzonitrile 97%
4-[(4-Chloro-2-pyrimidinyl)amino]benzonitrile
Formel:
C₁₁H₇ClN₄ Molecular Weight: 230,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443699 CAS-Nummer: 244768-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...