8-Acetyl-7-hydroxy-4-methylcoumarin

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Acetyl-4-methylumbelliferone

OR345062-5G
APOSOR345062-5GEA 383 EUR
APOSOR345062-5G
8-Acetyl-7-hydroxy-4-methylcoumarin
8-Acetyl-7-hydroxy-4-methylcoumarin
Formel: C₁₂H₁₀O₄
Molecular Weight: 218,21 g/mol
Schmelzpunkt: 170 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2555-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR