2-Amino-3,5-dichlor-4-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 3,5-Dichloro-4-methylpyridin-2-amine

OR345072-25G
APOSOR345072-25GEA 92.8 EUR
APOSOR345072-25G
2-Amino-3,5-dichlor-4-methylpyridin 97%
2-Amino-3,5-dichloro-4-methylpyridine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129031
CAS-Nummer: 31430-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR