6-Chloro-N-methyl-3-nitropyridin-2-amine 95+%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-5-nitro-6-methylaminopyridine
OR345079-1G
APOSOR345079-1GEA
109
EUR
APOSOR345079-1G
6-Chloro-N-methyl-3-nitropyridin-2-amine 95+%
6-Chloro-N-methyl-3-nitropyridin-2-amine
Formel:
C₆H₆ClN₃O₂ Molecular Weight: 187,5837 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460164 CAS-Nummer: 33742-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...