5-Bromo-6-hydroxy-2-methylpyridine 95%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-2-hydroxy-6-picoline

OR345086-1G
APOSOR345086-1GEA 71.2 EUR
APOSOR345086-1G
5-Bromo-6-hydroxy-2-methylpyridine 95%
5-Bromo-6-hydroxy-2-methylpyridine
Formel: C₆H₆BrNO
Molecular Weight: 188,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095217
CAS-Nummer: 374633-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR