5-Brom-3-iod-2-methoxypyridin 95%

Lieferant: Apollo Scientific

OR345114-1G
APOSOR345114-1GEA 72.5 EUR
APOSOR345114-1G
5-Brom-3-iod-2-methoxypyridin 95%
5-Brom-3-iod-2-methoxypyridin
Formel: C₆H₅BrINO
Lagertemperatur: Raumtemperatur
MDL: MFCD05663522
CAS-Nummer: 578007-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR