3-Hydroxy-2-nitrobenzoesäure
Lieferant: Apollo Scientific
OR345117-5G
APOSOR345117-5GEA
0
EUR
APOSOR345117-5G
3-Hydroxy-2-nitrobenzoesäure
3-Hydroxy-2-nitrobenzoic acid
Formel:
C₇H₅NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017003 CAS-Nummer: 602-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...