2-Brom-5-iod-3-methylpyridin 98%
Lieferant: Apollo Scientific
Synonyme:
2-Brom-5-iod-3-picolin
OR345123-5G
APOSOR345123-5GEA
77
EUR
APOSOR345123-5G
2-Brom-5-iod-3-methylpyridin 98%
2-Bromo-5-iodo-3-methylpyridine
Formel:
C₆H₅BrIN Molecular Weight: 297,92 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037466 CAS-Nummer: 65550-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...