2-(Benzyloxy)benzoesäure

Lieferant: Apollo Scientific

Synonyme: 2-Benzyloxybenzoic acid

OR345143-25G
APOSOR345143-25GEA 105 EUR
APOSOR345143-25G
2-(Benzyloxy)benzoesäure
2-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051940
CAS-Nummer: 14389-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR