5-Chloropyridine-3,4-diamine 97%
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-3,4-diaminopyridine
OR345157-1G
APOSOR345157-1GEA
104
EUR
APOSOR345157-1G
5-Chloropyridine-3,4-diamine 97%
5-Chloropyridine-3,4-diamine
Formel:
C5H6ClN3 Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263519 CAS-Nummer: 929074-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...