4-Methoxy-m-toluylsäure 98%

Lieferant: Apollo Scientific

OR345162-10G
APOSOR345162-10GEA 105 EUR
APOSOR345162-10G
4-Methoxy-m-toluylsäure 98%
4-Methoxy-m-toluylsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270114
CAS-Nummer: 6880-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR