4-Brom-2-chlor-5-methoxypyridin 95+%

Lieferant: Apollo Scientific

OR345169-250MG
APOSOR345169-250MGEA 46.4 EUR
APOSOR345169-250MG
4-Brom-2-chlor-5-methoxypyridin 95+%
4-Bromo-2-chloro-5-methoxypyridine
Formel: C₆H₅BrClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09972186
CAS-Nummer: 1020253-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR