6-Amino-3-brompicolinsäure 98%
Lieferant: Apollo Scientific
Synonyme:
6-Amino-3-bromo-2-pyridinecarboxylic acid
OR345173-1G
APOSOR345173-1GEA
104
EUR
APOSOR345173-1G
6-Amino-3-brompicolinsäure 98%
6-Amino-3-brompicolinsäure
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699642 CAS-Nummer: 1033201-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...