3-Amino-2-brom-6-chlorpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Brom-6-chlor-3-pyridylamin

OR345185-1G OR345185-5G
APOSOR345185-1GEA 0 EUR
APOSOR345185-1G APOSOR345185-5G
3-Amino-2-brom-6-chlorpyridin 97%
3-Amino-2-bromo-6-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11504927
CAS-Nummer: 1050501-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR