5-(Aminomethyl)-2-brompyridin 98%

Lieferant: Apollo Scientific
Achtung

OR345226-250MG
APOSOR345226-250MGEA 0 EUR
APOSOR345226-250MG
5-(Aminomethyl)-2-brompyridin 98%
5-(Aminomethyl)-2-brompyridin
Formel: C₆H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06738744
CAS-Nummer: 120740-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR