2,4-Dibrom-3-nitropyridin 97%

Lieferant: Apollo Scientific

OR345229-250MG
APOSOR345229-250MGEA 87 EUR
APOSOR345229-250MG
2,4-Dibrom-3-nitropyridin 97%
2,4-Dibromo-3-nitropyridine
Formel: C₅H₂Br₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09750299
CAS-Nummer: 121263-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR