3-Brom-5-nitropyridin 98%

Lieferant: Apollo Scientific

OR345270-5G
APOSOR345270-5GEA 60.9 EUR
APOSOR345270-5G
3-Brom-5-nitropyridin 98%
3-Bromo-5-nitropyridine
Formel: C₅H₃BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04114216
CAS-Nummer: 15862-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR