2,3-Dibrom-5-nitropyridin
Lieferant: Apollo Scientific
OR345271-5G
APOSOR345271-5GEA
68.3
EUR
APOSOR345271-5G
2,3-Dibrom-5-nitropyridin
2,3-Dibromo-5-nitropyridine
Formel:
C₅H₂Br₂N₂O₂ Molecular Weight: 281.8896 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233994 CAS-Nummer: 15862-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...