2-Cyanisonicotinsäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Cyano-4-pyridinecarboxylic acid
OR345273-5G
APOSOR345273-5GEA
105
EUR
APOSOR345273-5G
2-Cyanisonicotinsäure
2-Cyanoisonicotinic acid
Formel:
C₇H₄N₂O₂ Molecular Weight: 148,1200 g/mol Schmelzpunkt: 236 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411729 CAS-Nummer: 161233-97-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...