5-Benzyloxy-6-azaindole 95+%
Lieferant: Apollo Scientific
Synonyme:
5-(Benzyloxy)-1H-pyrrolo[2,3-c]pyridine
OR345279-250MG
APOSOR345279-250MGEA
183
EUR
APOSOR345279-250MG
5-Benzyloxy-6-azaindole 95+%
5-Benzyloxy-6-azaindole
Formel:
C₁₄H₁₂N₂O Molecular Weight: 224,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038914 CAS-Nummer: 17288-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...